![Anti-Cy3/Cy5 antibody, Mouse monoclonal clone CY-96, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/342/797/d2257bc6-464f-4ab0-9f01-510f334ba724/640/d2257bc6-464f-4ab0-9f01-510f334ba724.jpg)
Anti-Cy3/Cy5 antibody, Mouse monoclonal
![Anti-Cy3/Cy5 antibody, Mouse monoclonal clone CY-96, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/342/797/d2257bc6-464f-4ab0-9f01-510f334ba724/640/d2257bc6-464f-4ab0-9f01-510f334ba724.jpg)
Synonym(s):
Monoclonal Anti-Cy3/Cy5
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing