![Anti-COG5 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/565/566/e2998a23-10ea-4673-bb86-83fb1f8d8e05/640/e2998a23-10ea-4673-bb86-83fb1f8d8e05.jpg)
Anti-COG5 antibody produced in rabbit
![Anti-COG5 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/565/566/e2998a23-10ea-4673-bb86-83fb1f8d8e05/640/e2998a23-10ea-4673-bb86-83fb1f8d8e05.jpg)
Synonym(s):
Anti-13S Golgi transport complex 90 kDa subunit, Anti-COG complex subunit 5, Anti-Component of oligomeric Golgi complex 5, Anti-Conserved oligomeric Golgi complex subunit 5, Anti-GTC-90, Anti-Golgi transport complex 1
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing