Anti-Biotin−FITC antibody produced in goat
Synonym(s):
Anti-Biotin
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Conjugate
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing