Skip to Content
Merck
Ammonium tetraphenylborate 99%

Ammonium tetraphenylborate

Ammonium tetraphenylborate 99%
Synonym(s):
Tetraphenylboron ammonium
Linear Formula:
NH4B(C6H5)4
CAS Number:
Molecular Weight:
337.27

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing