Skip to Content
Merck
Aminobenzoate Sodium pharmaceutical secondary standard, certified reference material

Aminobenzoate sodium

Aminobenzoate Sodium pharmaceutical secondary standard, certified reference material
Synonym(s):
4-Aminobenzoic acid sodium salt, PABA, Sodium 4-aminobenzoate
Linear Formula:
C7H6NO2Na
CAS Number:
Molecular Weight:
159.12

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing