Skip to Content
Products
Cart
0
BE
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Aluminum-magnesium-silicon alloy, Al99Mg0.5Si0.5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF62748692
Aluminum-magnesium-silicon alloy, Al99Mg0.5Si0.5,
200mm rod, 5.0mm diameter
Expand
View Pricing
GF46655826
Aluminum-magnesium-silicon alloy, Al99Mg0.5Si0.5,
100mm rod, 5.0mm diameter
Expand
View Pricing
GF25179183
Aluminum-magnesium-silicon alloy, Al99Mg0.5Si0.5,
1m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF14577846
Aluminum-magnesium-silicon alloy, Al99Mg0.5Si0.5,
500mm rod, 5.0mm diameter
Expand
View Pricing
GF47397561
Aluminum-magnesium-silicon alloy, Al99Mg0.5Si0.5,
5m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF86877994
Aluminum-magnesium-silicon alloy, Al99Mg0.5Si0.5,
50m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF83067228
Aluminum-magnesium-silicon alloy, Al99Mg0.5Si0.5,
1000mm rod, 5.0mm diameter
Expand
View Pricing
GF24695028
Aluminum-magnesium-silicon alloy, Al99Mg0.5Si0.5,
25m wire, 0.25mm diameter, as drawn
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Français - FR
Learn More