
Alnustone

Synonym(s):
(4E,6E)-1,7-Diphenyl-4,6-heptadien-3-one, (4E,6E)-1,7-Diphenyl-4,6-heptadien-3-one
Empirical Formula (Hill Notation):
C19H18O
CAS Number:
Molecular Weight:
262.35
Products
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing