Skip to Content
Merck
Allethrin PESTANAL®, analytical standard

Allethrin

Allethrin PESTANAL®, analytical standard
Synonym(s):
Allylcinerine
Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing