Skip to Content
Merck
vibrant-m

Alitame

vibrant-m
Synonym(s):
L-α-Aspartyl-N-(2,2,4,4-tetramethyl-3-thietanyl)-D-alaninamide hemi(pentahydrate)
Empirical Formula (Hill Notation):
C14H25N3O4S · 2.5H2O
CAS Number:
Molecular Weight:
376.47

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing