![Aldolase from rabbit muscle ammonium sulfate suspension, 10-20 units/mg protein](/deepweb/assets/sigmaaldrich/product/images/286/028/b05bd63e-d8ef-4fb9-a864-0f6b29e7c4e3/640/b05bd63e-d8ef-4fb9-a864-0f6b29e7c4e3.jpg)
Aldolase from rabbit muscle
![Aldolase from rabbit muscle ammonium sulfate suspension, 10-20 units/mg protein](/deepweb/assets/sigmaaldrich/product/images/286/028/b05bd63e-d8ef-4fb9-a864-0f6b29e7c4e3/640/b05bd63e-d8ef-4fb9-a864-0f6b29e7c4e3.jpg)
Synonym(s):
D-Fructose-1,6-bisphosphate-D-glyceraldehyde-3-phosphate-lyase, Fructose-diphosphate Aldolase
CAS Number:
Products
Brand
Biological Source
Physical Form
Tissue
Chemical Composition
Quality Segment
Showing 1-2 of 2