Aegeline
Synonym(s):
Egeline
Empirical Formula (Hill Notation):
C18H19NO3
CAS Number:
Molecular Weight:
297.35
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing