Adenosine 5′-triphosphate disodium salt hydrate
Synonym(s):
ATP disodium salt, 5′-ATP-Na2, ATP disodium salt hydrate
Empirical Formula (Hill Notation):
C10H14N5Na2O13P3 · xH2O
Products
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-4 of 4
Compare
Product Number
Description
Pricing