Skip to Content
Merck
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate ≥98.0% (HPLC), powder

Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate

Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate ≥98.0% (HPLC), powder
Synonym(s):
3′,5′-Cyclic AMP sodium salt, cAMP-Na
Empirical Formula (Hill Notation):
C10H11N5NaO6P · H2O
CAS Number:
Molecular Weight:
369.20

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1