Skip to Content
Merck
Acycloguanosine ≥99% (HPLC), powder

Acycloguanosine

Acycloguanosine ≥99% (HPLC), powder
Synonym(s):
9-[(2-Hydroxyethoxy)methyl]guanine, Acyclovir
Empirical Formula (Hill Notation):
C8H11N5O3
CAS Number:
Molecular Weight:
225.20
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Activity Spectrum
Research Area
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing