Skip to Content
Merck
Acibenzolar-S-methyl PESTANAL®, analytical standard

Acibenzolar-S-methyl

Acibenzolar-S-methyl PESTANAL®, analytical standard
Synonym(s):
S-Methyl-1,2,3-benzothiadiazole-7-carbothioate
Empirical Formula (Hill Notation):
C8H6N2OS2
CAS Number:
Molecular Weight:
210.28
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2