![Acenaphthene-d10 99 atom % D](/deepweb/assets/sigmaaldrich/product/structures/337/577/ea7901fc-89a2-471e-b14a-3269685d0ffd/640/ea7901fc-89a2-471e-b14a-3269685d0ffd.png)
Acenaphthene-d10
![Acenaphthene-d10 99 atom % D](/deepweb/assets/sigmaaldrich/product/structures/337/577/ea7901fc-89a2-471e-b14a-3269685d0ffd/640/ea7901fc-89a2-471e-b14a-3269685d0ffd.png)
Synonym(s):
1,8-·Ethylenenaphthalene-D10, Deuterated acenaphthene
Empirical Formula (Hill Notation):
C12D10
CAS Number:
Molecular Weight:
164.27
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1