Acacetin
Synonym(s):
4′-Methylapigenin, 5,7-Dihydroxy-4′-methoxyflavone, Apigenin 4′-methyl ether, Buddleoflavonol, Linarigenin
Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing