![8-Bromotheophylline United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/201/332/c403ee46-2fe3-4fa6-a65c-f1b92bd6d7ed/640/c403ee46-2fe3-4fa6-a65c-f1b92bd6d7ed.png)
8-Bromotheophylline
![8-Bromotheophylline United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/201/332/c403ee46-2fe3-4fa6-a65c-f1b92bd6d7ed/640/c403ee46-2fe3-4fa6-a65c-f1b92bd6d7ed.png)
Synonym(s):
8-Bromo-3,9-dihydro-1,3-dimethyl-1H-purine-2,6-dione
Empirical Formula (Hill Notation):
C7H7BrN4O2
CAS Number:
Molecular Weight:
259.06
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing