![8arm-PEG10K-Vinylsulfone, tripentaerythritol core average Mn 10,000](/deepweb/assets/sigmaaldrich/product/structures/423/524/376fe7f4-20d7-4e85-8f75-49caad82201b/640/376fe7f4-20d7-4e85-8f75-49caad82201b.png)
8arm-PEG10K
![8arm-PEG10K-Vinylsulfone, tripentaerythritol core average Mn 10,000](/deepweb/assets/sigmaaldrich/product/structures/423/524/376fe7f4-20d7-4e85-8f75-49caad82201b/640/376fe7f4-20d7-4e85-8f75-49caad82201b.png)
Synonym(s):
8arm-PEG-Maleimide, 8arm-PEG-NH2, hexaglycerol core, 8arm-PEG-NH2, tripentaerythritol core, 8arm-PEG-SH, 8arm-PEG-Vinylsulfone, tripentaerythritol core
Linear Formula:
R(O(CH2CH2O)nCH2CH2OOSCHCH2)8
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Showing 1-5 of 5
Compare
Product Number
Description
Pricing