
6-Azido-hexanoic acid

Synonym(s):
6-Azido-hexanoic acid
Empirical Formula (Hill Notation):
C6H11N3O2
CAS Number:
Molecular Weight:
157.17
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Application
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing