Skip to Content
Merck
6-Amino-2H-1,4-benzoxazin-3(4H)-one 97%

6-Amino-2H-1,4-benzoxazin-3(4H)-one

6-Amino-2H-1,4-benzoxazin-3(4H)-one 97%
Empirical Formula (Hill Notation):
C8H8N2O2
CAS Number:
Molecular Weight:
164.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing