![5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt tablet](/deepweb/assets/sigmaaldrich/product/structures/524/334/1e302426-e770-4585-be58-eca3424b3be7/640/1e302426-e770-4585-be58-eca3424b3be7.png)
5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt
![5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt tablet](/deepweb/assets/sigmaaldrich/product/structures/524/334/1e302426-e770-4585-be58-eca3424b3be7/640/1e302426-e770-4585-be58-eca3424b3be7.png)
Synonym(s):
BC-Indicator, X-GlcA, X-glucuronide
Empirical Formula (Hill Notation):
C14H12BrClNNaO7
CAS Number:
Molecular Weight:
444.59
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing