![5-Bromo-3-indolyl β-D-galactopyranoside ≥98%](/deepweb/assets/sigmaaldrich/product/structures/176/870/92375fc5-fc85-49a8-a2a8-7f1ed61bcec3/640/92375fc5-fc85-49a8-a2a8-7f1ed61bcec3.png)
5-Bromo-3-indolyl β-D-galactopyranoside
![5-Bromo-3-indolyl β-D-galactopyranoside ≥98%](/deepweb/assets/sigmaaldrich/product/structures/176/870/92375fc5-fc85-49a8-a2a8-7f1ed61bcec3/640/92375fc5-fc85-49a8-a2a8-7f1ed61bcec3.png)
Synonym(s):
Bluo-Gal
Empirical Formula (Hill Notation):
C14H16BrNO6
CAS Number:
Molecular Weight:
374.18
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing