![4-tert-Octylphenol solution 1000 μg/mL in acetone, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/260/748/9b765e24-e603-4906-aecf-2bd0ef819e6d/640/9b765e24-e603-4906-aecf-2bd0ef819e6d.png)
4-tert-Octylphenol solution
![4-tert-Octylphenol solution 1000 μg/mL in acetone, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/260/748/9b765e24-e603-4906-aecf-2bd0ef819e6d/640/9b765e24-e603-4906-aecf-2bd0ef819e6d.png)
Synonym(s):
4-(1,1,3,3-Tetramethylbutyl)phenol solution
Empirical Formula (Hill Notation):
C14H22O
CAS Number:
Molecular Weight:
206.32
Products
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing