
4-Nitrophenyl stearate

Synonym(s):
4-Nitrophenyl octadecanoate, Octadecanoic acid 4-nitrophenyl ester
Empirical Formula (Hill Notation):
C24H39NO4
CAS Number:
Molecular Weight:
405.57
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1