Skip to Content
Merck
4-Nitrophenyl decanoate lipase substrate

4-Nitrophenyl decanoate

4-Nitrophenyl decanoate lipase substrate
Synonym(s):
4-Nitrophenyl caprate, Decanoic acid 4-nitrophenyl ester
Empirical Formula (Hill Notation):
C16H23NO4
CAS Number:
Molecular Weight:
293.36

Products

Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing