4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate
Synonym(s):
2-(Trimethylsilyl)ethyl 4-nitrophenyl carbonate, TEOC-ONp
Empirical Formula (Hill Notation):
C12H17NO5Si
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing