Skip to Content
Merck
4-Methyl-4H-1,2,4-triazole-3-thiol 97%

4-Methyl-4H-1,2,4-triazole-3-thiol

4-Methyl-4H-1,2,4-triazole-3-thiol 97%
Synonym(s):
3-Mercapto-4-methyl-4H-1,2,4-triazole
Empirical Formula (Hill Notation):
C3H5N3S
CAS Number:
Molecular Weight:
115.16
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing