![4-Hydroxyphenethyl bromide 96%](/deepweb/assets/sigmaaldrich/product/structures/253/628/b0f5f135-7070-49ce-ae5e-c4a0f2bf692a/640/b0f5f135-7070-49ce-ae5e-c4a0f2bf692a.png)
4-Hydroxyphenethyl bromide
![4-Hydroxyphenethyl bromide 96%](/deepweb/assets/sigmaaldrich/product/structures/253/628/b0f5f135-7070-49ce-ae5e-c4a0f2bf692a/640/b0f5f135-7070-49ce-ae5e-c4a0f2bf692a.png)
Synonym(s):
2-(4-Hydroxyphenyl)-1-bromoethane, 4-(2-Bromoethyl)phenol, 4-Hydroxy-1-(2-bromoethyl)benzene
Empirical Formula (Hill Notation):
C8H9BrO
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing