Skip to Content
Merck
4′-Formylbenzo-15-crown-5 97%

4′-Formylbenzo-15-crown-5

4′-Formylbenzo-15-crown-5 97%
Empirical Formula (Hill Notation):
C15H20O6
CAS Number:
Molecular Weight:
296.32

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing