4arm-PEG40K
Synonym(s):
4arm-PEG-Maleimide, 4arm-PEG-NH2, 4arm-PEG-Succinimidyl Carboxymethyl Ester
Linear Formula:
C(CH2O(CH2CH2O)nCH2COOC4H4O2N)4
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Showing 1-3 of 3