Skip to Content
Merck
4-Amino-2-(trifluoromethyl)benzonitrile 97%

4-Amino-2-(trifluoromethyl)benzonitrile

4-Amino-2-(trifluoromethyl)benzonitrile 97%
Synonym(s):
4-Cyano-3-trifluoromethylaniline
Linear Formula:
NCC6H3(CF3)NH2
CAS Number:
Molecular Weight:
186.13

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing