4,4′-Methylenebis(cyclohexylamine)
Synonym(s):
4,4′-Diaminodicyclohexylmethane
Linear Formula:
CH2(C6H10NH2)2
CAS Number:
Molecular Weight:
210.36
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Markush Class
Markush Group
Monomer Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing