Skip to Content
Merck
4-(4-Methoxybenzylamino)-7-nitrobenzofurazan

4-(4-Methoxybenzylamino)-7-nitrobenzofurazan

4-(4-Methoxybenzylamino)-7-nitrobenzofurazan
Synonym(s):
4-(4-Methoxybenzylamino)-7-nitro-2,1,3-benzoxadiazole, MBD
Empirical Formula (Hill Notation):
C14H12N4O4
CAS Number:
Molecular Weight:
300.27
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing