3-(Trimethylsilyl)-1-propanesulfonic acid-d6 sodium salt
Synonym(s):
2,2-Dimethyl-2-silapentane-5-sulfonate-d6 sodium salt, DSS-d6, Deuterated 3-(trimethylsilyl)-1-propanesulfonic acid sodium salt, Sodium 2-(trimethylsilyl)-1-propanesulfonate-d6
Linear Formula:
(CH3)3Si(CD2)3SO3Na
CAS Number:
Molecular Weight:
224.36
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing