
3-tert-Butyl-1-o-tolyl-1H-pyrazol-5-amine

Synonym(s):
3-tert-Butyl-1-(2-methylphenyl)-1H-pyrazol-5-amine
Empirical Formula (Hill Notation):
C14H19N3
CAS Number:
Molecular Weight:
229.32
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing