Skip to Content
Merck
3-(N,N-Dimethylmyristylammonio)propanesulfonate ≥98.0% (TLC)

3-(N,N-Dimethylmyristylammonio)propanesulfonate

3-(N,N-Dimethylmyristylammonio)propanesulfonate ≥98.0% (TLC)
Synonym(s):
3-(N,N-Dimethyltetradecylammonio)propanesulfonate, 3-(Myristyldimethylammonio)propanesulfonate, N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, Myristyl sulfobetaine, SB3-14
Linear Formula:
CH3(CH2)13N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
363.60
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Purity
Quality Segment
Application
Special Grade
Technique