![3-Nitrofluoranthene BCR®, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/334/111/a65edb9e-39ae-4078-8bb3-0276f24155ba/640/a65edb9e-39ae-4078-8bb3-0276f24155ba.png)
3-Nitrofluoranthene
![3-Nitrofluoranthene BCR®, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/334/111/a65edb9e-39ae-4078-8bb3-0276f24155ba/640/a65edb9e-39ae-4078-8bb3-0276f24155ba.png)
Empirical Formula (Hill Notation):
C16H9NO2
CAS Number:
Molecular Weight:
247.25
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing