Skip to Content
Merck
3-Methylenecyclobutanecarbonitrile 97% (GC)

3-Methylenecyclobutanecarbonitrile

3-Methylenecyclobutanecarbonitrile 97% (GC)
Synonym(s):
3-Methylene-1-cyanocyclobutane
Empirical Formula (Hill Notation):
C6H7N
CAS Number:
Molecular Weight:
93.13

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing