Skip to Content
Merck
3-Hydroxypropionitrile 97%

3-Hydroxypropionitrile

3-Hydroxypropionitrile 97%
Synonym(s):
2-Cyanoethanol, Ethylene cyanohydrin, Hydracrylonitrile, 3-Hydroxypropionitrile
Linear Formula:
HOCH2CH2CN
CAS Number:
Molecular Weight:
71.08

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-3 of 3
Compare
Product Number
Description
Pricing