Skip to Content
Merck
3-Chloro-1-phenyl-1-propyne 97%

3-Chloro-1-phenyl-1-propyne

3-Chloro-1-phenyl-1-propyne 97%
Synonym(s):
1-Chloro-3-phenyl-2-propyne, 3-Phenylpropargyl chloride
Empirical Formula (Hill Notation):
C9H7Cl
CAS Number:
Molecular Weight:
150.60

Products

Product Category
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing