Skip to Content
Merck
3-Amino-1-propanol analytical standard

3-Amino-1-propanol

3-Amino-1-propanol analytical standard
Synonym(s):
3-Aminopropyl alcohol, 3-Amino-1-propanol
Linear Formula:
HO(CH2)3NH2
CAS Number:
Molecular Weight:
75.11
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing