
3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt

Synonym(s):
3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt
Empirical Formula (Hill Notation):
C6H3Cl2NaO4S
CAS Number:
Molecular Weight:
265.05
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing