3,4-Thiophenedicarboxylic acid
Empirical Formula (Hill Notation):
C6H4O4S
CAS Number:
Molecular Weight:
172.16
Products
Product Category
Brand
Formula Weight
Melting Point (Ā°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing