![3,4-Dichloroisocoumarin serine protease inhibitor](/deepweb/assets/sigmaaldrich/product/structures/319/370/76c3e219-5355-4827-9a9a-9cb33c25dc1b/640/76c3e219-5355-4827-9a9a-9cb33c25dc1b.png)
3,4-Dichloroisocoumarin
![3,4-Dichloroisocoumarin serine protease inhibitor](/deepweb/assets/sigmaaldrich/product/structures/319/370/76c3e219-5355-4827-9a9a-9cb33c25dc1b/640/76c3e219-5355-4827-9a9a-9cb33c25dc1b.png)
Synonym(s):
3,4-DCI, 3,4-Dichloro-2-benzopyran-1-one, 3,4-Dichloroisocoumarin
Empirical Formula (Hill Notation):
C9H4Cl2O2
CAS Number:
Molecular Weight:
215.03
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing