Skip to Content
Merck
3,3′,5,5′-Tetramethylbenzidine dihydrochloride hydrate 97%

3,3′,5,5′-Tetramethylbenzidine dihydrochloride hydrate

3,3′,5,5′-Tetramethylbenzidine dihydrochloride hydrate 97%
Synonym(s):
TMB dihydrochloride
Linear Formula:
[-C6H2(CH3)2-4-NH2]2·2HCl·xH2O
CAS Number:
Molecular Weight:
313.27 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade