![3-(2-Iodoacetamido)-PROXYL free radical](/deepweb/assets/sigmaaldrich/product/structures/178/788/98c426db-e5a3-4382-87eb-7d86bd192088/640/98c426db-e5a3-4382-87eb-7d86bd192088.png)
3-(2-Iodoacetamido)-PROXYL
![3-(2-Iodoacetamido)-PROXYL free radical](/deepweb/assets/sigmaaldrich/product/structures/178/788/98c426db-e5a3-4382-87eb-7d86bd192088/640/98c426db-e5a3-4382-87eb-7d86bd192088.png)
Synonym(s):
3-(2-Iodoacetamido)-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, free radical
Empirical Formula (Hill Notation):
C10H18IN2O2
CAS Number:
Molecular Weight:
325.17
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing