Skip to Content
Merck
2-(Propylamino)ethanol ≥98%

2-(Propylamino)ethanol

2-(Propylamino)ethanol ≥98%
Synonym(s):
N-Propylethanolamine
Linear Formula:
CH3CH2CH2NHCH2CH2OH
CAS Number:
Molecular Weight:
103.16
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing