![2-Naphthoxyacetic acid 98%](/deepweb/assets/sigmaaldrich/product/structures/197/964/1f9b76ca-620a-4c1f-9bb8-ac6e3b58f900/640/1f9b76ca-620a-4c1f-9bb8-ac6e3b58f900.png)
2-Naphthoxyacetic acid
![2-Naphthoxyacetic acid 98%](/deepweb/assets/sigmaaldrich/product/structures/197/964/1f9b76ca-620a-4c1f-9bb8-ac6e3b58f900/640/1f9b76ca-620a-4c1f-9bb8-ac6e3b58f900.png)
Synonym(s):
BNOA, 2-Naphthoxyacetic acid
Linear Formula:
C10H7OCH2CO2H
CAS Number:
Molecular Weight:
202.21
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-3 of 3