Skip to Content
Merck
2-Methoxypyridine ≥98%

2-Methoxypyridine

2-Methoxypyridine ≥98%
Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing